Learn More
Medchemexpress LLC Thianthrene | 92-85-3 | MFCD00005065 | 100.0% | 216.33 g·mol⁻¹ | C12H8S2 | 10 G

Supplier: Medchemexpress LLC HYW01412010G
Thianthrene is a sulfur-containing tricyclic compound supplied for research use as a biochemical assay reagent and reported inhibitor of Leishmania donovani pteridine reductase 1 (PTR1). The product is offered as a solid and as a DMSO solution, with supporting documentation and safety data available for laboratory handling.
- High purity (≈99.98%).
- Molecular formula C12H8S2; molecular weight 216.33 g·mol⁻¹.
- Available as powder and as a 10 mM solution in DMSO.
- Offered in multiple pack sizes suitable for bench-scale work.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
- Includes product data sheet and SDS for safe laboratory use.
- Intended for research use only; not for human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.