Learn More
Medchemexpress LLC Thiazole bisamide (JNJ-61803534) | 1917306-14-9 | MFCD34676883 | 99.8% | 622.41 | C23H23Cl2F6N3O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1397805MG
Encompass_Preferred
JNJ-61803534 is a research compound identified as a potent, orally active RORγt inverse agonist with demonstrated anti-inflammatory activity, including inhibition of IL-17A production in human CD4+ T cells. It has been characterized with CAS 1917306-14-9, a defined molecular formula and weight, and is supplied for laboratory research use with supporting analytical data.
  • Potent RORγt inverse agonist with low-nanomolar activity.
  • Demonstrated inhibition of IL-17A production in human CD4+ T cells.
  • Characterized by molecular formula and molecular weight for identification.
  • Provided in quantified milligram-scale quantities suitable for in vitro studies.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-22717