Learn More
Apexbio Technology LLC Thiazole Orange 107091-89-4 250mg
SDP
Supplier:  Apexbio Technology LLC A4726250
Encompass_Preferred
Thiazole Orange (CAS 107091-89-4) is a fluorescent dye designed for nucleic acid detection Thiazole Orange exerts its biological activity primarily through intercalation and electrostatic interactions with double-stranded DNA or RNA resulting in enhanced fluorescence intensity In experimental assays it enables precise staining and visualization of nucleic acids Based on these pharmacological properties Thiazole Orange holds research potential in applications such as DNA quantification RNA localization analyses and hematological studies including the assessment of reticulocyte content in blood samples
Catalog No. 50-000-93541