Learn More
Medchemexpress LLC Thiazolidine-2,4-dicarboxylic acid | 30097-06-4 | MFCD00145399 | 99.8% | 177.18 g/mol | C5H7NO4S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0242411G
Encompass_Preferred
Tidiacic (thiazolidine-2,4-dicarboxylic acid) is a small-molecule research reagent reported to act as a hepatoprotective antioxidant and a sulfur donor. It is used in studies of liver protection, oxidative stress, and sulfur-transfer pathways. The compound is characterized by a low molecular weight and is supplied as a high-purity solid suitable for analytical and preparative research applications.
  • Acts as a hepatoprotective antioxidant and sulfur donor.
  • Useful for research into liver protection and oxidative-stress pathways.
  • Small-molecule with formula C5H7NO4S and molecular weight 177.18 g/mol.
  • High reported purity suitable for analytical applications.
Catalog No. 50-003-01605