Learn More
Medchemexpress LLC Thiazolidine, 3-[(2,3-dihydro-1H-inden-2-yl)acetyl]-4-(1-pyrrolidinylcarbonyl)-, (4R)- | 130849-58-0 | 99.7% | 344.47 g/mol | C19H24N2O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1912310MG
Encompass_Preferred
Z-321 is a small-molecule prolyl endopeptidase (PEP) inhibitor provided as a high-purity research reagent for use in in vitro and in vivo studies. Follow supplied safety, handling, and storage recommendations when preparing and using this compound.
  • High purity suitable for research applications.
  • Supplied with certificate of analysis and analytical data.
  • Provided as a stable powder for long-term storage.
  • Includes structural identifiers and SMILES for informatics use.
  • Recommended storage conditions extend shelf life and stability.
Catalog No. 50-002-95327