Learn More
Medchemexpress LLC Thiazolium,3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-5-(4,6,6-trihydroxy-4,6-dioxido-3,5-dioxa-4,6-diphosphahex-1-yl)-,chloride (1:1) | 154-87-0 | 99.9% | 460.77 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11307610MM/1ML
Encompass_Preferred
Thiamine pyrophosphate is the coenzyme form of Vitamin B1, acting as a required intermediate in several crucial metabolic complexes. It is essential for oxidative phosphorylation and the pentose phosphate pathway, functioning as a cofactor for α-ketoacid dehydrogenases.
  • Acts as a cofactor for α-ketoacid dehydrogenases
  • Inhibits ischemia-reperfusion induced oxidative damage
  • Prevents Cisplatin induced cardiotoxicity
  • Reduces associated oxidative stress and heart injury
Catalog No. 50-004-35599