Learn More
TARGETMOL CHEMICALS INC Thiazovivin 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T215510MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Thiazovivin, a ROCK inhibitor (IC50 0.5 uM), increases the survival rate of hESC. Purity 98.75%
Catalog No. 50-287-7681