Learn More
Medchemexpress LLC HY-13257 5mg Medchemexpress, Thiazovivin CAS:1226056-71-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY132575MG
Encompass_Preferred
Medchemexpress, HY-13257 5mg Thiazovivin CAS:1226056-71-8 Thiazovivin is a potent ROCK inhibitor, which can protect human embryonic stem cells. Thiazovivin improves the efficiency of iPSC generation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0266