Learn More
Selleck Chemical LLC Thiazovivin 5mg 1226056-71-8
SDP
Supplier:  Selleck Chemical LLC S14595MG
Encompass_Preferred
Thiazovivin is a novel ROCK inhibitor with IC50 of 0.5 ?M in a cell-free assay, promotes hESC survival after single-cell dissociation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-753-1