Learn More
PHYTOTECHNOLOGY LABS THIDIAZURON 1G
Supplier:  PHYTOTECHNOLOGY LABS T8881G
Encompass_Preferred
Thidiazuron (TDZ); >98% Purity, 51707-55-2, MFCD00078723, Formula C9H8N4OS, Formula Weight 220.25, Thidiazuron was originally used as a defoliant but this phenyl-urea also has cytokinin activity ,TDZ, 1-Phenyl-3-(1,2,3-thidiazol-5-yl)urea
Catalog No. NC1789769