Learn More
Bioworld Thidiazuron (TDZ), 100 mg
Supplier:  Bioworld 306310181
Encompass_Preferred
Thidiazuron (TDZ)Synonym: Phenyl-N'-(1,2,3-thiadiazol-5-yl) ureaCytokinin like growth regulator Absorbed via the leafPlant cell culture testedAppearance: white to off-white powderMelting Point: 210.5-212.5 ºCSoluble in DMSOIrritating to eyes, respiratory system and skin; do not breathe dustSolubilizeat 1 mg/mLin 0.1N KOH
Catalog No. 50-255-070