Learn More
Medchemexpress LLC Thieno[2,3-c]isoquinolin-5-one | 420849-22-5 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1217191MG
Encompass_Preferred
TIQ-A (Thieno[2,3-c]isoquinolin-5-one) is a small-molecule tankyrase (TNKS/poly-ADP-ribose polymerase) inhibitor supplied for research use. It shows potent activity against TNKS2 (IC50 = 24 nM) and has been reported as a potential anti-ischemic agent. The material is provided as a high-purity solid appropriate for biochemical and cellular studies.
  • Potent TNKS2 inhibitor with an IC50 of 24 nM.
  • High purity (99.85%) suitable for research assays.
  • Available in small research pack sizes for assay development.
  • Intended for biochemical and cellular mechanism-of-action studies (research use only).
Catalog No. 50-002-32469