Learn More
Medchemexpress LLC Thieno[2,3-c]pyridine-2-propanoic acid, 4-(4-cyanophenyl)-α,α-dimethyl- | 2013582-27-7 | 100.0% | 336.41 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1473005MG
Encompass_Preferred
Puliginurad (YL-90148) is a potent urate transporter (URAT) inhibitor. It can be used for hyperuricemia and gout research.
  • Potent urate transporter (URAT) inhibitor
  • Used for hyperuricemia and gout research
  • Molecular formula: C₁₉H₁₆N₂O₂S
  • Targets URAT1
  • Pathway: Membrane Transporter/Ion Channel
Catalog No. 50-003-60682