Learn More
Medchemexpress LLC Thienopyridone | 1018454-97-1 | 95.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12815310MG
Encompass_Preferred
Thienopyridone is a potent and selective phosphatase of regenerating liver (PRL) phosphatase inhibitor with IC50s of 173 nM, 277 nM and 128 nM for PRL-1, PRL-2, and PRL-3, respectively. It shows minimal effects on other phosphatases. Thienopyridone induces p130Cas cleavage and apoptosis and has anticancer effects. It is intended for research use only.
  • Potent and selective PRL phosphatase inhibitor
  • Minimal effects on other phosphatases
  • Induces p130Cas cleavage and apoptosis
  • Exhibits anticancer effects
  • Significantly inhibits tumor cell anchorage-independent growth
  • Suppresses HUVEC migration
Catalog No. 50-003-38414