Learn More
Cayman Chemical ThIenyldecyl IsothIocynAT 10mg
SDP
Supplier:  Cayman Chemical 1965310 MG
Encompass_Preferred
An antiproliferative agent that presumably modulates xenobiotic-metabolizing enzymes, such as inhibiting CYP450, and/or induces phase II detoxifying enzymes
Catalog No. 50-298-4772