Learn More
Medchemexpress LLC Thiethylperazine dimaleate | 1179-69-7 | 99.99% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB1794A5MG
Encompass_Preferred
Thiethylperazine dimaleate is a phenothiazine derivative, acting as an orally active and potent antagonist for dopamine D2-receptors and histamine H1-receptors. It also functions as a selective ABCC1 activator, contributing to the reduction of amyloid-β (Aβ) load in mice. This compound exhibits anti-emetic, antipsychotic, and antimicrobial effects.
  • Enhances antibiotic activity at low concentrations
  • Inhibits vancomycin-sensitive and resistant E. faecalis
  • Reduces Aβ42 levels in APP/PS1 mice in vivo
Catalog No. 50-004-12316