Learn More
Medchemexpress LLC Thifensulfuron-methyl | 79277-27-3 | MFCD00468118 | 98.0% | 387.39 g/mol | C12H13N5O6S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW020020100MG
Encompass_Preferred
Thifensulfuron-methyl is a sulfonylurea-class herbicide provided as a research and analytical standard. Supplied as a white to off-white solid, it has defined purity and storage recommendations to support reliable laboratory handling and herbicidal studies.
  • High purity (98.0%) suitable for analytical applications.
  • Solid powder form for convenient handling and weighing.
  • Well-characterized molecular formula and weight for accurate calculations.
  • Recommended storage conditions to maintain long-term stability.
  • Used to study control of broadleaved weeds in cereal and soybean crops.
  • Available in small pack sizes for research-scale experiments.
Catalog No. 50-003-01812