Learn More
Accuflow Systems Saint-Gobain Extraction Thimble, Ceramic, Coarse Porosity, 70 x 25 mm
Supplier:  Accuflow Systems 3254421
Encompass_Preferred
Coarse-porosity ceramic extraction thimbles designed for laboratory solvent extraction and residue ignition. Constructed from Alundum ceramic (Norton grade AN-485), these thimbles deliver rapid filtration, thermal durability, and easy cleaning by ignition. Typical applications include extraction of soaps, fats, foodstuffs, and rubber in analytical and preparative workflows.
  • Coarse porosity, retains particles ≈20 μm.
  • Alundum ceramic, Norton grade AN-485.
  • Dimensions: 70 x 25 mm (length x outer diameter).
  • Reusable; cleans by ignition for residue removal.
  • Suitable for solvent extraction of soaps, fats, foods, and rubber.
  • Available in pack quantities for laboratory use.
Catalog No. NC2754388