Learn More
Research Products International Corp Thimerosal, 10 Grams
SDP
Supplier:  Research Products International Corp T2104010.0
Encompass_Preferred
Bacteriostatic and fungistatic mercurial agent.
Toxicity: LD50 98 mg/kg
WARNING! This product can expose you to Thimerosal, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-213-683