Learn More
Research Products International Corp Thimerosal 500 G
SDP
Supplier:  Research Products International Corp T21040500.0
Encompass_Preferred
Bacteriostatic and fungistatic mercurial agent.
Toxicity: LD50 98 mg/kg
WARNING! This product can expose you to Thimerosal, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-223-0333