Learn More
Enzo Life Sciences Thimerosal (5g). CAS: 54-64-8
SDP
Supplier:  Enzo Life Sciences ALX400013G005
Encompass_Preferred
Modulates thiol groups in biological systems, especially in those involving cAMP, Ca2+ and inositol phosphate. Causes a release of calcium from intracellular stores in many cells types. Has toxic effects on several cell types. Induces apoptosis in vitro. Alternative name: Ethyl mercurithiosalicylic acid . Na, Mercurothiolate. Purity: ≥98% (HPLC). Solubility: Soluble in water (600mg/ml) or methanol. Long Term Storage: Ambient.
Catalog No. 50-200-9069