Learn More
Electron Microscopy Sciences Thin Sample Holder, 15 mm Pin
SDP
Catalog No. 5029307
Encompass
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For ISI, JEOL, TOPCON: double set screw for a secure holding of the specimen during observation. 15mm(dia). X 10mm(H), 6.4mm split
Catalog No. 50-293-07 Supplier Electron Microscopy Sciences Supplier No. 7594815
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals, etc. For ISI, JEOL, TOPCON: double set screw for a secure holding of the specimen during observation. 15mm(dia). X 10mm(H), 6.4mm split