Learn More
Andwin Scientific THIN STEM TRANSFER PIPETS; BUL
SDP
Supplier:  Andwin Scientific 10235500
Encompass_Preferred
With its polyethylene material, the thin stem transfer pipet features resistance against breaking or shattering. This product has a volume capacity of 4 mL and a bulb draw volume of 3.1 mL. The pipet has an overall length of 15.5 centimeters. It is sold in a pack of 500.
Catalog No. 50-819-431