Learn More
Electron Microscopy Sciences Veco Thin and Thick Bars 100+ym, Copper Grid
SDP
Supplier:  Electron Microscopy Sciences 100YMCU
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Hole: 100 µm
Catalog No. 50-980-161