Learn More
Medchemexpress LLC Thiobencarb | 28249-77-6 | 99.98% | 1 ML

Supplier: Medchemexpress LLC HY12979210MM/1ML
Thiobencarb is an herbicide that can reduce growth, photosynthetic activity, and the amount of Rieske iron-sulfur protein in the diatom Thalassiosira pseudonana. It is provided as a ready-to-use solution for laboratory applications.
- Purity: 99.98%
- Supplied as 1 mL of 10 mM solution in DMSO
- Storage at -20°C, protect from light
- In solvent storage at -80°C for 6 months or -20°C for 1 month (protect from light)
- CAS number: 28249-77-6
- Molecular weight: 257.79
- Chemical formula: C12H16ClNOS
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.