Learn More
Medchemexpress LLC Thiochrome | 92-35-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN724710MM/1ML
Encompass_Preferred
Thiochrome, a natural oxidation product and metabolite of thiamine, is a selective M4 muscarinic receptor of acetylcholine (ACh) affinity enhancer. It has neutral cooperativity with ACh at M1 to M3 receptors.
  • Natural oxidation product and metabolite of thiamine.
  • Selective M4 muscarinic receptor of acetylcholine (ACh) affinity enhancer.
  • Neutral cooperativity with ACh at M1 to M3 receptors.
  • Can increase the intensity of the reproduction process of one-cell organisms, worms, crustaceans, insects, and fishes.
Catalog No. 50-004-18016