Learn More
Medchemexpress LLC Thiochrome | 92-35-3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN724710MG
Encompass_Preferred
Thiochrome is a natural oxidation product and metabolite of thiamine. It acts as a selective M4 muscarinic receptor of acetylcholine (ACh) affinity enhancer and exhibits neutral cooperativity with ACh at M1 to M3 receptors.
  • Natural oxidation product and metabolite of thiamine
  • Selective M4 muscarinic receptor of acetylcholine (ACh) affinity enhancer
  • Exhibits neutral cooperativity with ACh at M1 to M3 receptors
  • Increases the intensity of reproduction in single-celled organisms, worms, crustaceans, insects, and fish
  • For research use only
Catalog No. 50-004-18547