Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Thiocolchicine-d3 10mg | 1314417-95-2 | 418.52 g·mol⁻1 | C22H22D3NO5S | 10 MG
SDP

Supplier:  Medchemexpress LLC HY116852S10MG

Encompass_Preferred

Thiocolchicine-d3 is a deuterium-labeled isotopologue of thiocolchicine used as an analytical standard and tracer in tubulin polymerization research, mass spectrometry, and quantitative assays.

  • Deuterium-labeled internal standard for quantitative analysis.
  • Purity ≥99.0%.
  • Molecular weight 418.52 g·mol⁻1.
  • Chemical formula C22H22D3NO5S.
  • Available in small package sizes for analytical workflows.

Catalog No. 50-002-28974


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.