Learn More
Cayman Chemical ThIodIglycolIc AcId-d4 1mg
SDP
Supplier:  Cayman Chemical 90024781 MG
Encompass_Preferred
An internal standard for the quantification of TDGA by GC- or LC-MS
Catalog No. 50-289-3427