Learn More
Cayman Chemical ThIodIglycolIc AcId-d4 500UG
SDP
Supplier:  Cayman Chemical 9002478500 UG
Encompass_Preferred
An internal standard for the quantification of TDGA by GC- or LC-MS
Catalog No. 50-289-4182