Learn More
Cayman Chemical ThIoFluor 623 10mg
SDP
Supplier:  Cayman Chemical 1308310 MG
Encompass_Preferred
Reacts with thiols resulting in an increase in fluorescence intensity of up to 120-fold; the response is selective for thiols, occurs in aqueous media, and results in a fluorophore with emission at 623 nm; cell-permeable
Catalog No. 50-295-3754