Learn More
Selleck Chemical LLC Thioguanine 50mg 154-42-7
SDP
Supplier:  Selleck Chemical LLC S177450MG
Encompass_Preferred
Thioguanine, a purine antimetabolite, inhibits DNMT1 activity through ubiquitin-targeted degradation, used in the treatment of acute lymphoblastic leukemia, autoimmune disorders (e.g., Crohn's disease, rheumatoid arthritis) and organ transplant recipients. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1621