Learn More
TRIBIOSCIENCE INC Thiol Fluorometric Assay 100 Tests
Supplier: TRIBIOSCIENCE INC TBS2026
Tribioscience’s Thiol Quantification Fluorometric Assay Kit provides a flexible, accurate, sensitive, and time-saving approach for detecting FREE thiol groups in a wide variety of samples.In this assay, the non-fluorescent probe binds to the thiol in the sample to produce a fluorescent product which can be measured at 400 nm excitation, 460 nm emission.