Learn More
Medchemexpress LLC Thiol-PEG-CH2COOH | 95.0% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140730100MG
Encompass_Preferred
Thiol-PEG-CH2COOH (MW 5000) is a PEG-based PROTAC linker used in the synthesis of PROTACs. These PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
  • Solid appearance
  • White to off-white color
  • Average molecular weight of 5000
Catalog No. 50-003-55101