Learn More
Biotium Thiol Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF740 1/EA
Supplier:  Biotium 96109
Encompass_Preferred
CF Dye Succinimidyl Esters (SE also known as NHS esters) are Biotiums next-generation fluorescent dyes in amine-reactive form for labeling antibodies and other proteins on amine groups such as lysine residues Near-infrared fluorescent CF740 has excitation/emission at 742/767 nm
Catalog No. 50-005-01454