Learn More
Biotium Thiol Reactive Dyes Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF790 1/EA
Supplier:  Biotium 96108
Encompass_Preferred
CF Dye Maleimides are thiol-reactive fluorescent dyes Maleimides are commonly used to label proteins on free thiol groups CF Dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes Near-IR fluorescent CF790 dye has excitation/emission at 783/808 nm
Catalog No. 50-005-01453