Learn More
Abcam Thiolutin, antibiotic agent, 5MG
Supplier:  Abcam AB1435565MG
Encompass_Preferred
MW 228.3 Da, Purity >98%. Sulfur-based antibiotic agent. Potent RNA polymerase inhibitor. Inhibits endothelial cell adhesion (IC₅₀ = 0.83 μM). Suppresses tumor cell induced angiogenesis and shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6110