Learn More
Medchemexpress LLC Thiomichler's ketone (4,4'-bis(dimethylamino)thiobenzophenone) | 1226-46-6 | 81.1% | 284.42 | 500 MG
Supplier: Medchemexpress LLC HYW009311500MG
Thiomichler's ketone is a biological material or organic compound primarily used in life science research. It functions as a photometric reagent for the determination of various metals, including gold, palladium, platinum, mercury, silver, and copper.
- Functions as a photometric reagent
- Useful for the determination of gold, palladium, platinum, mercury, silver, and copper
- Widely used in life science research
- Solid appearance
- Color: brown to reddish brown