Learn More
Medchemexpress LLC Thiomorpholine | 123-90-0 | 103.19 | 25 G

Supplier: Medchemexpress LLC 25GHYW007359
Thiomorpholine is a biochemical reagent used as a biological material or organic compound for life science related research. It can be utilized to synthesize quinolines when coupled with bromide and can form polymer derivatives studied as oxidative sensitive and hemocompatible materials.
- Used as a biological material or organic compound for life science related research
- Can be used to synthesize quinolines when coupled with bromide
- Can form polymer derivatives that are studied as oxidative sensitive materials
- Can form polymer derivatives that are studied as hemocompatible materials
- Appearance: liquid
- Color: colorless to light yellow
- Density: 1.026 g/cm³
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.