Learn More
Cayman Chemical ThIomuscImol 5mg
SDP
Supplier:  Cayman Chemical 188575 MG
Encompass_Preferred
A GABAA receptor agonist (IC50 = 19 nM) that has been used as a photoaffinity label for the purification and identification of GABA binding sites within the GABAA receptor complex
Catalog No. 50-298-4004