Learn More
TARGETMOL CHEMICALS INC THIOMYRISTOYL 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T344710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Thiomyristoyl is an effective and selective SIRT2 inhibitor (IC50 28 nM). It inhibits SIRT1 (IC50 98 uM) but no effect on SIRT3 even at 200 uM. purity: 99%
Catalog No. 50-257-6942