Learn More
Medchemexpress LLC Thiophene-3-carboxylic acid | 88-13-1 | 128.15 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW004615
Encompass_Preferred
Thiophene-3-carboxylic acid is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It has been used in the preparation of novel acetylene monomer, required for the synthesis of poly(2-methylbut-2-enyl thiophene-3-carboxylate) and oxazolines and oxazoles. It is also used as a leading compound for the development of a clinically useful D-amino acid inhibitor and has the potential to serve as active site probes to elucidate the structure-function relationships of D-amino acids.
  • Used as a biochemical reagent for life science research.
  • Can be used as a biological material or organic compound.
  • Prepares novel acetylene monomer for various syntheses.
  • Serves as a leading compound for D-amino acid inhibitor development.
  • Potential for elucidating structure-function relationships of D-amino acids.
Catalog No. 50-004-48085