Learn More
Cayman Chemical ThIophnate-methyl 100mg
SDP
Supplier:  Cayman Chemical 35402100 MG
Encompass_Preferred
A carbamate fungicide; active against a variety of fungi (EC50s = <1 to 1-10 µM); induces hepatotoxicity and increases hepatic ROS in zebrafish larvae and adults at 12.5 and 25 mg/L
Catalog No. 50-291-3728