Learn More
Sigma Organic Chemistry Thiophosphoryl chloride | 1KG | 3982-91-0 | MFCD00011504
Supplier: Sigma Organic Chemistry 2242941.4KG
CAS #: 3982-91-0
MDL #: MFCD00011504
Purity: >98 %
UNSPSC Code: 12352100
Molecular Weight: 169.4
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.