Learn More
Medchemexpress LLC Thioridazine-d3 hydrochloride | 1189928-36-6 | 98.25% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB0965AS5MG
Encompass_Preferred
Thioridazine-d3 hydrochloride is a deuterium-labeled Thioridazine, acting as an antagonist of dopamine receptor D2 family proteins with potent antipsychotic and anti-anxiety activities. It also inhibits PI3K-Akt-mTOR signaling pathways, exhibiting an anti-angiogenic effect, and induces antiproliferative and apoptosis effects in various cancer cells, specifically targeting cancer stem cells (CSCs).
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS methods
Catalog No. 50-004-09100