Learn More
Medchemexpress LLC Thioridazine hydrochloride | 130-61-0 | 100.0% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB096510MM/1ML
Encompass_Preferred
Thioridazine hydrochloride is an orally active antagonist of dopamine receptor D2 family proteins, exhibiting potent anti-psychotic and anti-anxiety activities. It also acts as a potent inhibitor of PI3K-Akt-mTOR signaling pathways, demonstrating anti-angiogenic effects. This compound shows antiproliferative and apoptosis induction effects in various types of cancer cells, specifically targeting cancer stem cells (CSCs).
  • Orally active dopamine receptor D2 antagonist
  • Potent anti-psychotic and anti-anxiety agent
  • Inhibits PI3K-Akt-mTOR signaling pathways
  • Exhibits anti-angiogenic effects
  • Induces antiproliferative and apoptosis in cancer cells
  • Targets cancer stem cells
  • Soluble in water (100 mg/mL) and DMSO (≥ 45 mg/mL)
  • Stable at 4°C in sealed storage
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C
Catalog No. 50-004-07979