Learn More
Enzo Life Sciences Thiorphan (5mg). CAS: 76721-89-6
SDP
Supplier:  Enzo Life Sciences BMLPI1130005
Encompass_Preferred
Inhibits the metalloprotease neprilysin (neutral endopeptidase 24.11) and neprilysin 2. Inhibits opioid-degrading enzymes, potentiating THC-induced antinociception. Potentiates the effects of natriuretic peptides. Blocks enzymatic degradation of luteinizing hormone releasing hormone (LHRH). Purity: ≥97% (TLC). Long Term Storage: -20°C.
Catalog No. 50-201-1450