Learn More
Medchemexpress LLC Thiosulfuric acid (H2S2O3), sodium salt (1:2) | 7772-98-7 | 99.9% | 158.11 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY108775A50G
Encompass_Preferred
Sodium thiosulfate is an antioxidant. It inhibits the expression of p-GSK-3β and β-catenin proteins, reduces IL-1β, COX-2, and Iba-1, and inhibits NFκB activation.
  • Promotes angiogenesis.
  • Inhibits inflammation.
  • Improves acute lung injury.
  • Exhibits anti-cancer activity against melanoma.
  • Exerts renal protective effects.
  • Can be used in the research of osteoarthritis, brain inflammation, cancer (such as breast cancer, melanoma), and kidney disease.
Catalog No. 50-003-46057