Learn More
Medchemexpress LLC Thiotaurine | 2937-54-4 | 141.21 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN151561MG
Encompass_Preferred
Thiotaurine is a metabolite of Cystine and acts as a sulfur donor. It is believed to modulate GAPDH activity, participating in neutrophil activation and leukocyte energy metabolism. Thiotaurine is recognized as a potential anti-inflammatory agent and immunomodulator.
  • Acts as a sulfur donor
  • May modulate GAPDH activity
  • Participates in neutrophil activation
  • Participates in leukocyte energy metabolism
  • Potential anti-inflammatory agent
  • Immunomodulator
  • Endogenous metabolite
Catalog No. 50-004-15675