Learn More
Medchemexpress LLC Thiotaurine | 2937-54-4 | 141.21 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN15156100MG
Encompass_Preferred
Thiotaurine, a metabolite of Cystine, acts as a sulfur donor and may modulate GAPDH activity, influencing neutrophil activation and leukocyte energy metabolism. It is recognized as a potential anti-inflammatory agent and immunomodulator.
  • Metabolite of Cystine
  • Functions as a sulfur donor
  • Modulates GAPDH activity
  • Involved in neutrophil activation
  • Contributes to leukocyte energy metabolism
  • Potential anti-inflammatory agent
  • Acts as an immunomodulator
Catalog No. 50-004-15632